Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
我們的服務對象
生物技術
生物製藥
CDMO
實驗室診斷
工業與應用科學
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh07231581_m1
See other CELF2 GE Assays ›
Gene Symbol:
CELF2
Gene Name:
CUGBP Elav-like family member 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 11603734 - 12468374 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh07231581_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2,900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2,900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2,900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
新增到購物車
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: CELF2
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015146431.2
XP_015001917.1
1284
103
XM_015146432.2
XP_015001918.1
1578
103
XM_015146434.2
XP_015001920.1
1457
103
XM_015146436.2
XP_015001922.1
1584
103
XM_015146437.2
XP_015001923.1
1541
103
XM_015146438.2
XP_015001924.1
1566
103
XM_015146443.2
XP_015001929.1
1663
103
XM_015146444.2
XP_015001930.1
1657
103
XM_015146445.2
XP_015001931.1
1645
103
XM_015146446.2
XP_015001932.1
1363
103
XM_028825955.1
XP_028681788.1
1234
103
XM_028825956.1
XP_028681789.1
1842
103
XM_028825957.1
XP_028681790.1
1824
103
XM_028825958.1
XP_028681791.1
1488
103
XM_028825959.1
XP_028681792.1
1457
103
XM_028825960.1
XP_028681793.1
1807
103
XM_028825961.1
XP_028681794.1
1293
103
XM_028825962.1
XP_028681795.1
1807
103
XM_028825963.1
XP_028681796.1
1597
103
XM_028825964.1
XP_028681797.1
1458
103
XM_028825965.1
XP_028681798.1
1458
103
XM_028825966.1
XP_028681799.1
1566
103
XM_028825967.1
XP_028681800.1
1405
103
XM_028825968.1
XP_028681801.1
1675
103
XM_028825969.1
XP_028681802.1
1455
103
XM_028825970.1
XP_028681803.1
1534
103
XM_028825971.1
XP_028681804.1
1476
103
XM_028825972.1
XP_028681805.1
1412
103
XM_028825973.1
XP_028681806.1
1258
103
XM_028825974.1
XP_028681807.1
1399
103
XM_028825975.1
XP_028681808.1
1329
103
XM_028825976.1
XP_028681809.1
1346
103
XM_028825977.1
XP_028681810.1
1442
103
XM_028825978.1
XP_028681811.1
1645
103
XM_028825979.1
XP_028681812.1
1498
103
XM_028825980.1
XP_028681813.1
1357
103
XM_028825981.1
XP_028681814.1
1461
103
XM_028825982.1
XP_028681815.1
1124
103
GenBank mRNA
JU332566.1
-
1284
103
JU332567.1
-
1360
103
JU332568.1
-
1342
103
JU332569.1
-
1342
103
JU474039.1
-
1482
103
JV489861.1
-
1365
103
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Macaca mulatta
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline