Search
Search

GBGT1| Gene Symbol: GBGT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028834664.1 | XP_028690497.1 | 179 | 79 | ||
| XM_028834666.1 | XP_028690499.1 | 179 | 79 | |||
| XM_028834670.1 | XP_028690503.1 | 178 | 79 | |||
| GenBank mRNA | JV540914.1 | - | 70 | 79 | ||
| JV612969.1 | - | 130 | 79 | |||
Set Membership: |
|