Search
Search

FRY| Gene Symbol: FRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837192.1 | XP_028693025.1 | 949 | 94 | ||
| XM_028837194.1 | XP_028693027.1 | 569 | 94 | |||
| XM_028837195.1 | XP_028693028.1 | 569 | 94 | |||
| XM_028837196.1 | XP_028693029.1 | 91 | 94 | |||
| GenBank mRNA | JU332415.1 | - | 95 | 94 | ||
| JU332416.1 | - | 95 | 94 | |||
| JV602975.1 | - | 398 | 94 | |||
| JV704899.1 | - | 151 | 94 | |||
Set Membership: |
|