Search
Search

B4GALT2| Gene Symbol: B4GALT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137763.2 | XP_014993249.1 | 1332 | 73 | ||
| XM_028834538.1 | XP_028690371.1 | 1128 | 73 | |||
| GenBank mRNA | JV524214.1 | - | 985 | 73 | ||
| JV632097.1 | - | 1192 | 73 | |||
Set Membership: |
|