Search
Search

FAT3| Gene Symbol: FAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084587.4 | XP_001084587.3 | 10659 | 72 | ||
| XM_015115461.2 | XP_014970947.2 | 10728 | 72 | |||
| XM_015115462.2 | XP_014970948.2 | 10659 | 72 | |||
| XM_015115463.2 | XP_014970949.2 | 10728 | 72 | |||
| XM_028833904.1 | XP_028689737.1 | 10950 | 72 | |||
| XM_028833905.1 | XP_028689738.1 | 10669 | 72 | |||
| GenBank mRNA | JV706571.1 | - | 1437 | 72 | ||
Set Membership: |
|