Search
Search

DGKH| Gene Symbol: DGKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120972.2 | XP_014976458.2 | 1777 | 73 | ||
| XM_015120975.2 | XP_014976461.1 | 1979 | 73 | |||
| XM_015120976.2 | XP_014976462.1 | 3813 | 73 | |||
| XM_015120978.2 | XP_014976464.1 | 906 | 73 | |||
| XM_028837488.1 | XP_028693321.1 | 1742 | 73 | |||
| GenBank mRNA | JV708149.1 | - | 1581 | 73 | ||
Set Membership: |
|