Search
Search

MIER1| Gene Symbol: MIER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141830.2 | XP_014997316.1 | 983 | 91 | ||
| XM_015141845.2 | XP_014997331.1 | 874 | 91 | |||
| XM_015141849.2 | XP_014997335.1 | 1089 | 91 | |||
| XM_015141853.2 | XP_014997339.1 | 1057 | 91 | |||
| XM_015141859.2 | XP_014997345.1 | 1109 | 91 | |||
| XM_015141867.2 | XP_014997353.1 | 1112 | 91 | |||
| XM_015141876.2 | XP_014997362.1 | 1116 | 91 | |||
| XM_015141884.2 | XP_014997370.1 | 1000 | 91 | |||
| XM_015141889.2 | XP_014997375.1 | 1131 | 91 | |||
| XM_015141894.2 | XP_014997380.1 | 1074 | 91 | |||
| XM_015141904.2 | XP_014997390.1 | 908 | 91 | |||
| XM_015141915.2 | XP_014997401.1 | 874 | 91 | |||
| XM_028832757.1 | XP_028688590.1 | 1075 | 91 | |||
| GenBank mRNA | JU320719.1 | - | 868 | 91 | ||
| JU320720.1 | - | 958 | 91 | |||
| JU476575.1 | - | 1082 | 91 | |||
| JU476576.1 | - | 895 | 91 | |||
| JV045415.1 | - | 879 | 91 | |||
Set Membership: |
|