Search
Search

TTC37| Gene Symbol: TTC37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015140451.2 | XP_014995937.1 | 1307 | 81 | ||
| XM_028849576.1 | XP_028705409.1 | 1016 | 81 | |||
| XM_028849577.1 | XP_028705410.1 | 1307 | 81 | |||
| XM_028849578.1 | XP_028705411.1 | 314 | 81 | |||
| GenBank mRNA | JU322825.1 | - | 1268 | 81 | ||
| JU477965.1 | - | 1246 | 81 | |||
| JV048805.1 | - | 1251 | 81 | |||
| JV488034.1 | - | 1138 | 81 | |||
| JV565212.1 | - | 1242 | 81 | |||
| JV727588.1 | - | 1282 | 81 | |||