Search
Search

NFATC3| Gene Symbol: NFATC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126454.2 | XP_014981940.1 | 2082 | 110 | ||
| XM_015126455.2 | XP_014981941.1 | 2083 | 110 | |||
| XM_028841545.1 | XP_028697378.1 | 2084 | 110 | |||
| XM_028841546.1 | XP_028697379.1 | 1994 | 110 | |||
| GenBank mRNA | JU334667.1 | - | 1793 | 110 | ||
| JU475714.1 | - | 1865 | 110 | |||
| JU475715.1 | - | 3782 | 110 | |||
Set Membership: |
|