Search
Search

SLC35F5| Gene Symbol: SLC35F5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015109948.2 | XP_014965434.1 | 533 | 114 | ||
| XM_015109950.2 | XP_014965436.1 | 128 | 114 | |||
| XM_015109951.2 | XP_014965437.1 | 533 | 114 | |||
| XM_028830646.1 | XP_028686479.1 | 131 | 114 | |||
| XM_028830647.1 | XP_028686480.1 | 131 | 114 | |||
| XM_028830648.1 | XP_028686481.1 | 131 | 114 | |||
| XM_028830649.1 | XP_028686482.1 | 131 | 114 | |||
| XM_028830651.1 | XP_028686484.1 | 924 | 114 | |||
| GenBank mRNA | JU336495.1 | - | 293 | 114 | ||
| JU493994.1 | - | 401 | 114 | |||
| JV048308.1 | - | 310 | 114 | |||
Set Membership: |
|