Search
Search

TCAIM| Gene Symbol: TCAIM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131203.2 | XP_014986689.2 | 1206 | 91 | ||
| XM_015131204.2 | XP_014986690.2 | 936 | 91 | |||
| XM_028844010.1 | XP_028699843.1 | 760 | 91 | |||
| XM_028844011.1 | XP_028699844.1 | 1209 | 91 | |||
| GenBank mRNA | JU475247.1 | - | 818 | 91 | ||
| JV712791.1 | - | 919 | 91 | |||