Search
Search

FOCAD| Gene Symbol: FOCAD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028835128.1 | XP_028690961.1 | 3086 | 97 | ||
| XM_028835129.1 | XP_028690962.1 | 3045 | 97 | |||
| XM_028835130.1 | XP_028690963.1 | 3102 | 97 | |||
| GenBank mRNA | JU322806.1 | - | 2996 | 97 | ||
| JU470716.1 | - | 2985 | 97 | |||
| JV511099.1 | - | 2901 | 97 | |||