Search
Search

PISD| Gene Symbol: PISD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150084.2 | XP_015005570.1 | 942 | 98 | ||
| XM_015150086.2 | XP_015005572.1 | 505 | 98 | |||
| XM_015150088.2 | XP_015005574.1 | 568 | 98 | |||
| XM_028827807.1 | XP_028683640.1 | 929 | 98 | |||
| XM_028827808.1 | XP_028683641.1 | 727 | 98 | |||
| XM_028827810.1 | XP_028683643.1 | 1237 | 98 | |||
| XM_028827811.1 | XP_028683644.1 | 1207 | 98 | |||
| XM_028827812.1 | XP_028683645.1 | 790 | 98 | |||
| XM_028827813.1 | XP_028683646.1 | 958 | 98 | |||
| XM_028827814.1 | XP_028683647.1 | 961 | 98 | |||
| XM_028827815.1 | XP_028683648.1 | 747 | 98 | |||
| XM_028827816.1 | XP_028683649.1 | 1789 | 98 | |||
| XM_028827817.1 | XP_028683650.1 | 544 | 98 | |||
| XM_028827818.1 | XP_028683651.1 | 510 | 98 | |||
| XM_028827819.1 | XP_028683652.1 | 771 | 98 | |||
| XM_028827820.1 | XP_028683653.1 | 774 | 98 | |||
| XM_028827821.1 | XP_028683654.1 | 929 | 98 | |||
| XM_028827823.1 | XP_028683656.1 | 359 | 98 | |||
| XM_028827824.1 | XP_028683657.1 | 599 | 98 | |||
| XM_028827825.1 | XP_028683658.1 | 339 | 98 | |||
| GenBank mRNA | JU333755.1 | - | 428 | 98 | ||
| JU475000.1 | - | 416 | 98 | |||
| JV046630.1 | - | 420 | 98 | |||
| JV463120.1 | - | 104 | 98 | |||
Set Membership: |
|