Search
Search

NCOA7| Gene Symbol: NCOA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015137488.2 | XP_014992974.1 | 5293 | 101 | ||
| XM_015137489.2 | XP_014992975.1 | 5494 | 101 | |||
| XM_015137490.2 | XP_014992976.1 | 4963 | 101 | |||
| XM_015137491.2 | XP_014992977.1 | 5065 | 101 | |||
| XM_015137492.2 | XP_014992978.1 | 5279 | 101 | |||
| XM_015137494.2 | XP_014992980.1 | 5199 | 101 | |||
| XM_015137495.2 | XP_014992981.1 | 5025 | 101 | |||
| XM_015137496.2 | XP_014992982.1 | 5745 | 101 | |||
| XM_028847073.1 | XP_028702906.1 | 5024 | 101 | |||
| XM_028847074.1 | XP_028702907.1 | 5007 | 101 | |||
| XM_028847075.1 | XP_028702908.1 | 5275 | 101 | |||
| GenBank mRNA | JV673286.1 | - | 518 | 101 | ||
Set Membership: |
|