Search
Search

ZNF430| Gene Symbol: ZNF430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002801148.3 | XP_002801194.1 | 179 | 104 | ||
| XM_015123416.2 | XP_014978902.1 | 179 | 104 | |||
| GenBank mRNA | JU324877.1 | - | 159 | 104 | ||
| JU494005.1 | - | 159 | 104 | |||
Set Membership: |
|