Search
Search

CELSR2| Gene Symbol: CELSR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147640.2 | XP_015003126.2 | 3692 | 129 | ||
| XM_028830686.1 | XP_028686519.1 | 3218 | 129 | |||
| XM_028830687.1 | XP_028686520.1 | 3219 | 129 | |||
| XM_028830691.1 | XP_028686524.1 | 3217 | 129 | |||
| GenBank mRNA | JV708777.1 | - | 3120 | 129 | ||
Set Membership: |
|