Search
Search

UTS2B| Gene Symbol: UTS2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015132510.2 | XP_014987996.1 | 212 | 76 | ||
| XM_028843562.1 | XP_028699395.1 | 212 | 76 | |||
| XM_028843563.1 | XP_028699396.1 | 212 | 76 | |||
Set Membership: |
|