Search
Search

C3H7orf57| Gene Symbol: C3H7orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133571.2 | XP_014989057.2 | 983 | 59 | ||
| XM_015133572.2 | XP_014989058.2 | 935 | 59 | |||
| XM_015133573.2 | XP_014989059.2 | 1056 | 59 | |||
| XM_028845782.1 | XP_028701615.1 | 1007 | 59 | |||
Set Membership: |
|