Search
Search

EBF3| Gene Symbol: EBF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002805861.3 | XP_002805907.1 | 1102 | 88 | ||
| XM_015148455.2 | XP_015003941.1 | 1102 | 88 | |||
| XM_015148458.2 | XP_015003944.1 | 1155 | 88 | |||
| XM_028827008.1 | XP_028682841.1 | 1635 | 88 | |||
| XM_028827009.1 | XP_028682842.1 | 1635 | 88 | |||
| XM_028827010.1 | XP_028682843.1 | 1129 | 88 | |||
| XM_028827011.1 | XP_028682844.1 | 1635 | 88 | |||
| XM_028827012.1 | XP_028682845.1 | 1635 | 88 | |||
| XM_028827013.1 | XP_028682846.1 | 1350 | 88 | |||
| XM_028827014.1 | XP_028682847.1 | 1635 | 88 | |||
Set Membership: |
|