Search
Search

KCP| Gene Symbol: KCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028846264.1 | XP_028702097.1 | 2691 | 112 | ||
| XM_028846265.1 | XP_028702098.1 | 2696 | 112 | |||
| XM_028846266.1 | XP_028702099.1 | 2690 | 112 | |||
| XM_028846267.1 | XP_028702100.1 | 2696 | 112 | |||
| XM_028846268.1 | XP_028702101.1 | 2695 | 112 | |||
| XM_028846269.1 | XP_028702102.1 | 2695 | 112 | |||
| XM_028846270.1 | XP_028702103.1 | 2693 | 112 | |||
Set Membership: |
|