Search
Search

MYPN| Gene Symbol: MYPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015147514.2 | XP_015003000.2 | 2726 | 87 | ||
| XM_015147515.2 | XP_015003001.2 | 3503 | 87 | |||
| XM_028826595.1 | XP_028682428.1 | 3981 | 87 | |||
| XM_028826596.1 | XP_028682429.1 | 3059 | 87 | |||
Set Membership: |
|