Search
Search

TSNAXIP1| Gene Symbol: TSNAXIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001094646.4 | XP_001094646.3 | 1770 | 79 | ||
| XM_001094769.4 | XP_001094769.3 | 1977 | 79 | |||
| XM_015126432.2 | XP_014981918.1 | 1737 | 79 | |||
| XM_015126433.2 | XP_014981919.1 | 525 | 79 | |||
| XM_028841531.1 | XP_028697364.1 | 1977 | 79 | |||
| XM_028841532.1 | XP_028697365.1 | 1643 | 79 | |||