Search
Search

C6H5orf58| Gene Symbol: C6H5orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141388.2 | XP_014996874.1 | 451 | 142 | ||
| XM_015141389.2 | XP_014996875.1 | 425 | 142 | |||
| XM_015141390.2 | XP_014996876.1 | 455 | 142 | |||
Set Membership: |
|