Search
Search

PPDPF| Gene Symbol: PPDPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266949.1 | NP_001253878.1 | 127 | 77 | ||
| GenBank mRNA | JU323102.1 | - | 123 | 77 | ||
| JU473566.1 | - | 59 | 77 | |||
| JV045483.1 | - | 127 | 77 | |||
| JV638295.1 | - | 128 | 77 | |||
| JV704077.1 | - | 124 | 77 | |||
Set Membership: |
|