Search
Search

KPNA2| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266111.2 | NP_001253040.1 | 100 | 71 | ||
| GenBank mRNA | JU334436.1 | - | 96 | 71 | ||
| JU475525.1 | - | 103 | 71 | |||
| JV047009.1 | - | 102 | 71 | |||
| JV047010.1 | - | 102 | 71 | |||
| JV047011.1 | - | 102 | 71 | |||
Set Membership: |
|