Search
Search
| Gene Symbol: Prkcz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022507.1 | NP_071952.1 | 13 - 14 | 1429 | 109 | |
| XM_006239535.2 | XP_006239597.1 | 10 - 11 | 3924 | 109 | ||
| XM_006239536.2 | XP_006239598.1 | 10 - 11 | 3130 | 109 | ||
| XM_008764337.1 | XP_008762559.1 | 13 - 14 | 1330 | 109 | ||
| GenBank mRNA | J04532.1 | - | 13 - 14 | 1429 | 109 | |
| M18332.1 | - | 10 - 11 | 982 | 109 | ||
| U27191.1 | - | 10 - 11 | 1194 | 109 | ||
Set Membership: |
|