Search
Search

Pafah1b1| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031763.3 | NP_113951.1 | 1169 | 88 | ||
| GenBank mRNA | AF016049.1 | - | 984 | 88 | ||
| BC072510.1 | - | 1169 | 7104733 | 88 | ||
Set Membership: |
|