Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00458026_m1
          See other MKNK2 GE Assays ›
          Gene Symbol:
          Mknk2
          Gene Name:
          MAP kinase-interacting serine/threonine kinase 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 11908060 - 11919163 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00458026_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mknk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011985.1 NP_001011985.1 5 - 6 484 62
          XM_006240883.2 XP_006240945.1 5 - 6 573 62
          XM_006240884.1 XP_006240946.1 4 - 5 461 62
          GenBank mRNA BC085941.1 - 5 - 6 484 7124839 62
          FQ211808.1 - 5 - 6 527 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000188.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mknk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of translation
          protein phosphorylation
          hemopoiesis
          intracellular signal transduction
          cellular response to arsenic-containing substance
          extrinsic apoptotic signaling pathway in absence of ligand
          protein serine/threonine kinase activity
          ATP binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P019215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 930037 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264900 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352757 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272945 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713971 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019215 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P019215 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P019215 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 44684 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10893754 - -
          Affymetrix Rat Gene 1.0 ST Array 10893752 - -
          Affymetrix Rat Gene 1.0 ST Array 10893755 - -
          Affymetrix Rat Gene 1.0 ST Array 10893745 - -
          Affymetrix Rat Gene 1.0 ST Array 10893750 - -
          Affymetrix Rat Gene 1.0 ST Array 10893753 - -
          Affymetrix Rat Gene 1.0 ST Array 10893746 - -
          Affymetrix Rat Gene 1.0 ST Array 10893751 - -
          Affymetrix Rat Gene 1.0 ST Array 10893757 - -
          Affymetrix Rat Gene 1.0 ST Array 10893749 - -
          Affymetrix Rat Gene 1.0 ST Array 10893756 - -
          Affymetrix Rat Gene 1.0 ST Array 10893748 - -
          Affymetrix Rat Gene 1.0 ST Array 10893747 - -
          Affymetrix Rat Gene 1.0 ST Array 10893758 - -
          Affymetrix Rat Gene 1.1 ST Array 10893744 - -
          Rat Genome 230 2.0 Array 1372038_PM_at - -
          Rat Genome 230 2.0 Array 1372617_PM_at - -
          Rat Genome 230 2.0 Array 1372617_at - -
          Rat Genome 230 2.0 Array 1372038_at - -
          Affymetrix HT Rat Focus Array 1372038_PM_at - -
          Affymetrix HT Rat Focus Array 1372038_at - -
          Affymetrix Rat Clariom S Assay PSR0700001604.rn.1 TC0700000188.rn.1 RTA TC
          Rat Expression Set 230 1372038_at - -
          Rat Expression Set 230 1372038_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10893744 - -
          Rat Genome U34 Set rc_AI137941_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21658529 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1376280 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline