Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00504906_m1
          See other ERG GE Assays ›
          Gene Symbol:
          Erg
          Gene Name:
          v-ets avian erythroblastosis virus E26 oncogene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 35645317 - 35749643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00504906_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Erg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133397.2 NP_596888.1 8 - 9 906 89
          XM_006248142.2 XP_006248204.1 9 - 10 1137 89
          XM_006248143.2 XP_006248205.1 8 - 9 1068 89
          XM_006248145.2 XP_006248207.1 7 - 8 995 89
          GenBank mRNA AB031088.1 - 8 - 9 906 89
          BC072519.1 - 8 - 9 886 7105647 89
          FQ217228.1 - 8 - 9 925 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1100001372.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Erg

          Panther Classification:

          Molecular Function -

          winged helix/forkhead transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell differentiation
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P095972 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P322881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265137 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155505 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 886904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916675 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28063 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212649 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646084 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 797522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2542 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 116488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466727 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941377 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683220 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107208 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973478 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45663 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 777692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704580 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 486029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 346550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191119 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 136346 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 10208 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23046 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 64299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39495 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 89191 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180274 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46579 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P322881 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P095972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P322881 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P322881 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P095972 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P525876 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52128 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79820 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78242 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 84007 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83683 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 94055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10753281 - -
          Affymetrix Rat Gene 1.0 ST Array 10753280 - -
          Affymetrix Rat Gene 1.0 ST Array 10753285 - -
          Affymetrix Rat Gene 1.0 ST Array 10753291 - -
          Affymetrix Rat Gene 1.0 ST Array 10753284 - -
          Affymetrix Rat Gene 1.0 ST Array 10753283 - -
          Affymetrix Rat Gene 1.0 ST Array 10753289 - -
          Affymetrix Rat Gene 1.0 ST Array 10753287 - -
          Affymetrix Rat Gene 1.0 ST Array 10753290 - -
          Affymetrix Rat Gene 1.0 ST Array 10753282 - -
          Affymetrix Rat Gene 1.0 ST Array 10753288 - -
          Affymetrix Rat Gene 1.1 ST Array 10753279 - -
          Rat Genome 230 2.0 Array 1387789_PM_at - -
          Rat Genome 230 2.0 Array 1387789_at - -
          Affymetrix HT Rat Focus Array 1387789_at - -
          Affymetrix HT Rat Focus Array 1387789_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1100008184.rn.1 TC1100001372.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008183.rn.1 TC1100001372.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1100008185.rn.1 TC1100001372.rn.1 RTA TC
          Rat Expression Set 230 1387789_PM_at - -
          Rat Expression Set 230 1387789_at - -
          Affymetrix Rat Gene 1.0 ST Array 10753279 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20841265 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367872 - -
          RatRef-12 Expression BeadChip ILMN_48833 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline