Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561490_m1
          See other MBL1 GE Assays ›
          Gene Symbol:
          Mbl1
          Gene Name:
          mannose-binding lectin (protein A) 1
          Gene Aliases:
          Mbpa, Mlb1
          Chromosome Location:
          Chr.16: 18736154 - 18742193 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561490_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mbl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012599.2 NP_036731.2 3 - 4 338 75
          GenBank mRNA BC088159.1 - 3 - 4 338 7326244 75
          FQ209936.1 - 3 - 4 354 75
          FQ218582.1 - 3 - 4 366 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000220.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mbl1

          Panther Classification:

          Molecular Function -

          surfactant

          Gene Ontology Categories:

          Function(s) Process(es)

          complement activation, lectin pathway
          complement activation, classical pathway
          negative regulation of growth of symbiont in host
          positive regulation of phagocytosis
          defense response to Gram-positive bacterium
          protein homotetramerization
          killing by host of symbiont cells
          protein homotrimerization
          protease binding
          calcium ion binding
          protein binding
          mannose binding
          carbohydrate binding
          polysaccharide binding
          protein homodimerization activity
          calcium-dependent protein binding
          phosphatidylinositol-4-phosphate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P228059 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 522491 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P228059 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P228059 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P228059 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P228059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 52615 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10787110 - -
          Affymetrix Rat Gene 1.0 ST Array 10787109 - -
          Affymetrix Rat Gene 1.0 ST Array 10787112 - -
          Affymetrix Rat Gene 1.0 ST Array 10787111 - -
          Affymetrix Rat Gene 1.0 ST Array 10787108 - -
          Affymetrix Rat Gene 1.1 ST Array 10787107 - -
          Rat Genome 230 2.0 Array 1387765_PM_at - -
          Rat Genome 230 2.0 Array 1387765_at - -
          Affymetrix HT Rat Focus Array 1387765_at - -
          Affymetrix HT Rat Focus Array 1387765_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600002118.rn.1 TC1600000220.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1600002115.rn.1 TC1600000220.rn.1 RTA TC
          Rat Expression Set 230 1387765_at - -
          Rat Expression Set 230 1387765_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10787107 - -
          Rat Genome U34 Set AF080507_at - -
          Rat Genome U34 Set AF080507_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21588453 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_2039269 - -
          RatRef-12 Expression BeadChip ILMN_59501 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline