Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00562472_m1
          See other GPD2 GE Assays ›
          Gene Symbol:
          Gpd2
          Gene Name:
          glycerol-3-phosphate dehydrogenase 2, mitochondrial
          Gene Aliases:
          mtGPDH
          Chromosome Location:
          Chr.3: 43223927 - 43355860 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00562472_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpd2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012736.1 NP_036868.1 16 - 17 2148 120
          GenBank mRNA BC083565.1 - 16 - 17 2106 7134834 120
          FQ223746.1 - 2404 120
          U08027.1 - 16 - 17 2127 120
          X78593.1 - 16 - 17 2148 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300000617.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpd2

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          glycerol-3-phosphate metabolic process
          gluconeogenesis
          NADH metabolic process
          glycerol catabolic process
          multicellular organism growth
          camera-type eye development
          oxidation-reduction process
          glycerol-3-phosphate dehydrogenase [NAD+] activity
          calcium ion binding
          sn-glycerol-3-phosphate:ubiquinone-8 oxidoreductase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P068809 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P021676 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42186 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689641 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 108556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44821 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 845180 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704284 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729054 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 550756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 379270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 855006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821515 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 853855 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 133699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 541583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 972203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 839824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 319937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 79621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765691 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 639368 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91144 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 31779 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 4339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 939281 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 336833 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 264869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 158957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245571 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394304 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462635 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 327775 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737353 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195999 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69816 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 128426 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76776 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90734 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 127093 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1819 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 90932 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 87192 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117892 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 1331 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 161033 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174441 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 157749 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78174 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021676 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068809 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P068809 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P021676 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P068809 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P021676 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P158899 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8126 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 34742 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32703 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 89596 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2244 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 63404 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96431 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53580 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90453 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10836279 - -
          Affymetrix Rat Gene 1.0 ST Array 10836290 - -
          Affymetrix Rat Gene 1.0 ST Array 10836285 - -
          Affymetrix Rat Gene 1.0 ST Array 10836281 - -
          Affymetrix Rat Gene 1.0 ST Array 10836282 - -
          Affymetrix Rat Gene 1.0 ST Array 10836292 - -
          Affymetrix Rat Gene 1.0 ST Array 10836284 - -
          Affymetrix Rat Gene 1.0 ST Array 10836294 - -
          Affymetrix Rat Gene 1.0 ST Array 10836286 - -
          Affymetrix Rat Gene 1.0 ST Array 10836295 - -
          Affymetrix Rat Gene 1.0 ST Array 10836289 - -
          Affymetrix Rat Gene 1.0 ST Array 10836278 - -
          Affymetrix Rat Gene 1.0 ST Array 10836283 - -
          Affymetrix Rat Gene 1.0 ST Array 10836288 - -
          Affymetrix Rat Gene 1.0 ST Array 10836287 - -
          Affymetrix Rat Gene 1.0 ST Array 10836296 - -
          Affymetrix Rat Gene 1.0 ST Array 10836291 - -
          Affymetrix Rat Gene 1.0 ST Array 10836280 - -
          Affymetrix Rat Gene 1.0 ST Array 10836293 - -
          Affymetrix Rat Gene 1.1 ST Array 10836277 - -
          Rat Genome 230 2.0 Array 1369666_at - -
          Rat Genome 230 2.0 Array 1387670_at - -
          Rat Genome 230 2.0 Array 1369666_PM_at - -
          Rat Genome 230 2.0 Array 1387670_PM_at - -
          Affymetrix HT Rat Focus Array 1369666_PM_at - -
          Affymetrix HT Rat Focus Array 1369666_at - -
          Affymetrix HT Rat Focus Array 1387670_at - -
          Affymetrix HT Rat Focus Array 1387670_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0300004781.rn.1 TC0300000617.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300004780.rn.1 TC0300000617.rn.1 RTA TC
          Rat Expression Set 230 1369666_PM_at - -
          Rat Expression Set 230 1387670_at - -
          Rat Expression Set 230 1387670_PM_at - -
          Rat Expression Set 230 1369666_at - -
          Affymetrix Rat Gene 1.0 ST Array 10836277 - -
          Rat Genome U34 Set U83880UTR#1_at - -
          Rat Genome U34 Set X78593_at - -
          Rat Genome U34 Set X78593_g_at - -
          Rat Genome U34 Set U83880UTR#1_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20718129 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366549 - -
          RatRef-12 Expression BeadChip ILMN_65127 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline