Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564087_m1
          See other EEF2K GE Assays ›
          Gene Symbol:
          Eef2k
          Gene Name:
          eukaryotic elongation factor-2 kinase
          Gene Aliases:
          SMEF2K
          Chromosome Location:
          Chr.1: 190780211 - 190839926 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564087_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eef2k
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012947.2 NP_037079.1 9 - 10 1226 64
          XM_006230156.2 XP_006230218.1 10 - 11 1550 64
          XM_006230157.2 XP_006230219.1 10 - 11 1947 64
          XM_006230158.2 XP_006230220.1 10 - 11 2627 64
          GenBank mRNA BC061825.1 - 10 - 11 1481 5599075 64
          U93849.1 - 9 - 10 1226 64
          X96426.1 - 1516 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002508.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eef2k

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of protein autophosphorylation
          protein autophosphorylation
          elongation factor-2 kinase activity
          calcium ion binding
          calmodulin binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P517591 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P086610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 728396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 514397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 429849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756888 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 425714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473564 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45498 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506990 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 391672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578133 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286547 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454435 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 732207 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721482 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343976 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149019 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168964 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85996 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123101 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086610 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P517591 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P086610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P086610 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P517591 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P155816 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P517591 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700603988 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700274659 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24458 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7537 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24564 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 71178 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16623 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23018 - -
          Agilent Rat cDNA Microarray (G4105A) 700274659 - -
          Agilent Rat cDNA Microarray (G4105A) 700603988 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10710504 - -
          Affymetrix Rat Gene 1.0 ST Array 10710512 - -
          Affymetrix Rat Gene 1.0 ST Array 10710495 - -
          Affymetrix Rat Gene 1.0 ST Array 10710497 - -
          Affymetrix Rat Gene 1.0 ST Array 10710500 - -
          Affymetrix Rat Gene 1.0 ST Array 10710510 - -
          Affymetrix Rat Gene 1.0 ST Array 10710509 - -
          Affymetrix Rat Gene 1.0 ST Array 10710502 - -
          Affymetrix Rat Gene 1.0 ST Array 10710505 - -
          Affymetrix Rat Gene 1.0 ST Array 10710507 - -
          Affymetrix Rat Gene 1.0 ST Array 10710498 - -
          Affymetrix Rat Gene 1.0 ST Array 10710508 - -
          Affymetrix Rat Gene 1.0 ST Array 10710506 - -
          Affymetrix Rat Gene 1.0 ST Array 10710503 - -
          Affymetrix Rat Gene 1.0 ST Array 10710511 - -
          Affymetrix Rat Gene 1.0 ST Array 10710496 - -
          Affymetrix Rat Gene 1.0 ST Array 10710499 - -
          Affymetrix Rat Gene 1.0 ST Array 10710501 - -
          Affymetrix Rat Gene 1.1 ST Array 10710494 - -
          Rat Genome 230 2.0 Array 1369638_at - -
          Rat Genome 230 2.0 Array 1394253_at - -
          Rat Genome 230 2.0 Array 1389999_at - -
          Rat Genome 230 2.0 Array 1387658_PM_at - -
          Rat Genome 230 2.0 Array 1387658_at - -
          Rat Genome 230 2.0 Array 1394253_PM_at - -
          Rat Genome 230 2.0 Array 1369638_PM_at - -
          Rat Genome 230 2.0 Array 1389999_PM_at - -
          Affymetrix HT Rat Focus Array 1387658_PM_at - -
          Affymetrix HT Rat Focus Array 1389999_at - -
          Affymetrix HT Rat Focus Array 1394253_PM_at - -
          Affymetrix HT Rat Focus Array 1394253_at - -
          Affymetrix HT Rat Focus Array 1389999_PM_at - -
          Affymetrix HT Rat Focus Array 1369638_at - -
          Affymetrix HT Rat Focus Array 1387658_at - -
          Affymetrix HT Rat Focus Array 1369638_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100019304.rn.1 TC0100002508.rn.1 RTA TC
          Rat Expression Set 230 1389999_PM_at - -
          Rat Expression Set 230 1387658_PM_at - -
          Rat Expression Set 230 1394253_at - -
          Rat Expression Set 230 1387658_at - -
          Rat Expression Set 230 1394253_PM_at - -
          Rat Expression Set 230 1389999_at - -
          Rat Expression Set 230 1369638_at - -
          Rat Expression Set 230 1369638_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10710494 - -
          Rat Genome U34 Set X96426_at - -
          Rat Genome U34 Set rc_AI059692_s_at - -
          Rat Genome U34 Set rc_AI102667_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21495989 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366045 - -
          RatRef-12 Expression BeadChip ILMN_61025 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline