Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00564852_m1
          See other TSC22D1 GE Assays ›
          Gene Symbol:
          Tsc22d1
          Gene Name:
          TSC22 domain family, member 1
          Gene Aliases:
          TS22A, TSC-22, Tgfb1i4
          Chromosome Location:
          Chr.15: 58554358 - 58658156 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00564852_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tsc22d1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109912.1 NP_001103382.1 2 - 3 3229 89
          NM_013043.2 NP_037175.1 2 - 3 357 89
          XM_006252339.1 XP_006252401.1 3 - 4 3101 89
          GenBank mRNA BC059146.1 - 2 - 3 368 89
          BC127547.1 - 2 - 3 349 89
          CB771530.1 - 2 - 3 357 89
          L25785.1 - 2 - 3 350 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1500000862.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tsc22d1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of cell proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          transcription factor activity, sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P14977 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P365516 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856083 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375203 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 643455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 836579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494238 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289412 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 311754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653001 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 526456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11822 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 222729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344832 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504014 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 620568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186634 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 26248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 646262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 369587 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 617707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 606783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 927358 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347257 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 893904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 819974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168200 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962160 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460835 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929876 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843687 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84206 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19944 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 139484 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105842 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 49909 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54513 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75506 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106700 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142176 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 43920 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 86847 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14977 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P365516 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P14977 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P365516 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P14977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P14977 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P365516 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700783673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38235 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58958 - -
          Agilent Rat cDNA Microarray (G4105A) 700783673 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10781585 - -
          Affymetrix Rat Gene 1.0 ST Array 10781590 - -
          Affymetrix Rat Gene 1.0 ST Array 10781588 - -
          Affymetrix Rat Gene 1.0 ST Array 10781583 - -
          Affymetrix Rat Gene 1.0 ST Array 10781582 - -
          Affymetrix Rat Gene 1.0 ST Array 10781586 - -
          Affymetrix Rat Gene 1.0 ST Array 10781591 - -
          Affymetrix Rat Gene 1.0 ST Array 10781589 - -
          Affymetrix Rat Gene 1.0 ST Array 10781587 - -
          Affymetrix Rat Gene 1.0 ST Array 10781584 - -
          Affymetrix Rat Gene 1.1 ST Array 10781581 - -
          Rat Genome 230 2.0 Array 1392438_PM_at - -
          Rat Genome 230 2.0 Array 1398759_PM_at - -
          Rat Genome 230 2.0 Array 1392438_at - -
          Rat Genome 230 2.0 Array 1398759_at - -
          Rat Genome 230 2.0 Array 1381257_PM_at - -
          Rat Genome 230 2.0 Array 1381257_at - -
          Affymetrix HT Rat Focus Array 1381257_PM_at - -
          Affymetrix HT Rat Focus Array 1392438_at - -
          Affymetrix HT Rat Focus Array 1398759_PM_at - -
          Affymetrix HT Rat Focus Array 1381257_at - -
          Affymetrix HT Rat Focus Array 1398759_at - -
          Affymetrix HT Rat Focus Array 1392438_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1500005961.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005981.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005963.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005962.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005978.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005979.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005980.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005964.rn.1 TC1500000862.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1500005965.rn.1 TC1500000862.rn.1 RTA TC
          Rat Expression Set 230 1398759_at - -
          Rat Expression Set 230 1398759_PM_at - -
          Rat Expression Set 230 1392438_PM_at - -
          Rat Expression Set 230 1381257_at - -
          Rat Expression Set 230 1381257_PM_at - -
          Rat Expression Set 230 1392438_at - -
          Affymetrix Rat Gene 1.0 ST Array 10781581 - -
          Rat Genome U34 Set L25785_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21194676 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366057 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline