Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00566386_g1
          See other BGLAP GE Assays ›
          Gene Symbol:
          Bglap
          Gene Name:
          bone gamma-carboxyglutamate (gla) protein
          Gene Aliases:
          Bglap2, Bgp, Bgpr, Bgpra
          Chromosome Location:
          Chr.2: 187741770 - 187748446 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00566386_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Bglap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013414.1 NP_038200.1 3 - 4 222 104
          XM_006232594.2 XP_006232656.1 4 - 5 3279 104
          GenBank mRNA M11777.1 - 3 - 4 202 104
          X04141.1 - 3 - 4 221 104

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200004033.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Bglap

          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          osteoblast differentiation
          osteoblast development
          aging
          cell aging
          response to mechanical stimulus
          response to gravity
          response to inorganic substance
          response to zinc ion
          response to organic cyclic compound
          response to activity
          regulation of bone mineralization
          biomineral tissue development
          response to nutrient levels
          response to vitamin K
          response to vitamin D
          response to testosterone
          response to hydroxyisoflavone
          odontogenesis
          response to drug
          response to estrogen
          response to ethanol
          response to glucocorticoid
          bone development
          cellular response to vitamin D
          cellular response to growth factor stimulus
          cellular response to BMP stimulus
          calcium ion binding
          structural constituent of bone

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898654 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15447 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15447 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10824293 - -
          Affymetrix Rat Gene 1.0 ST Array 10824291 - -
          Affymetrix Rat Gene 1.0 ST Array 10824292 - -
          Affymetrix Rat Gene 1.0 ST Array 10824290 - -
          Affymetrix Rat Gene 1.1 ST Array 10824289 - -
          Rat Genome 230 2.0 Array 1369142_PM_at - -
          Rat Genome 230 2.0 Array 1369142_at - -
          Affymetrix HT Rat Focus Array 1369142_at - -
          Affymetrix HT Rat Focus Array 1369142_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0200027270.rn.1 TC0200004033.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027267.rn.1 TC0200004033.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027268.rn.1 TC0200004033.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200027269.rn.1 TC0200004033.rn.1 RTA TC
          Rat Expression Set 230 1369142_PM_at - -
          Rat Expression Set 230 1369142_at - -
          Affymetrix Rat Gene 1.0 ST Array 10824289 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20768678 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1365188 - -
          RatRef-12 Expression BeadChip ILMN_56543 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline