Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00567994_m1
          See other MECR GE Assays ›
          Gene Symbol:
          Mecr
          Gene Name:
          mitochondrial trans-2-enoyl-CoA reductase
          Gene Aliases:
          Nrbf1
          Chromosome Location:
          Chr.5: 150001263 - 150027407 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00567994_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mecr
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017209.1 NP_058905.1 1 - 2 199 85
          GenBank mRNA AB015724.1 - 1 - 2 199 85
          FQ210707.1 - 1 - 2 195 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0500001585.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mecr

          Panther Classification:

          Molecular Function -

          reductase

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          fatty acid biosynthetic process
          oxidation-reduction process
          receptor binding
          zinc ion binding
          trans-2-enoyl-CoA reductase (NADPH) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11860 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 864425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 28538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682592 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1069 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382319 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781603 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 483823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263514 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 580237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489440 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828181 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18955 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 31420 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11860 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11860 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11860 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11860 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700856827 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13059 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79786 - -
          Agilent Rat cDNA Microarray (G4105A) 700856827 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10872521 - -
          Affymetrix Rat Gene 1.0 ST Array 10872516 - -
          Affymetrix Rat Gene 1.0 ST Array 10872512 - -
          Affymetrix Rat Gene 1.0 ST Array 10872514 - -
          Affymetrix Rat Gene 1.0 ST Array 10872520 - -
          Affymetrix Rat Gene 1.0 ST Array 10872517 - -
          Affymetrix Rat Gene 1.0 ST Array 10872518 - -
          Affymetrix Rat Gene 1.0 ST Array 10872519 - -
          Affymetrix Rat Gene 1.0 ST Array 10872515 - -
          Affymetrix Rat Gene 1.0 ST Array 10872513 - -
          Affymetrix Rat Gene 1.1 ST Array 10872511 - -
          Rat Genome 230 2.0 Array 1398252_PM_at - -
          Rat Genome 230 2.0 Array 1398252_at - -
          Affymetrix HT Rat Focus Array 1398252_PM_at - -
          Affymetrix HT Rat Focus Array 1398252_at - -
          Affymetrix Rat Clariom S Assay PSR0500011396.rn.1 TC0500001585.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011393.rn.1 TC0500001585.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0500011392.rn.1 TC0500001585.rn.1 RTA TC
          Rat Expression Set 230 1398252_at - -
          Rat Expression Set 230 1398252_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10872511 - -
          Rat Genome U34 Set AB015724_at - -
          Rat Genome U34 Set AB015724_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22152688 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1350310 - -
          RatRef-12 Expression BeadChip ILMN_68668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline