Search
Search
| Gene Symbol: Gabrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017289.1 | NP_058985.1 | 1 - 2 | 116 | 75 | |
| GenBank mRNA | BC087714.1 | - | 1 - 2 | 81 | 7317154 | 75 |
| M35162.1 | - | 1 - 2 | 116 | 75 | ||
Set Membership: |
|