Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00569900_m1
          See other SMAD2 GE Assays ›
          Gene Symbol:
          Smad2
          Gene Name:
          SMAD family member 2
          Gene Aliases:
          Madh2
          Chromosome Location:
          Chr.18: 72550105 - 72612078 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00569900_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001277450.1 NP_001264379.1 6 - 7 942 67
          NM_019191.2 NP_062064.1 6 - 7 970 67
          XM_006254945.2 XP_006255007.1 5 - 6 966 67
          XM_006254946.2 XP_006255008.1 6 - 7 683 67
          GenBank mRNA AB010147.1 - 5 - 6 744 67
          AB017912.1 - 6 - 7 836 67
          AF056001.1 - 6 - 7 870 67
          BC127497.1 - 6 - 7 814 8364718 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad2

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          in utero embryonic development
          endoderm formation
          mesoderm formation
          transcription, DNA-templated
          protein phosphorylation
          transforming growth factor beta receptor signaling pathway
          common-partner SMAD protein phosphorylation
          SMAD protein complex assembly
          zygotic specification of dorsal/ventral axis
          gastrulation
          pattern specification process
          endoderm development
          heart development
          negative regulation of cell proliferation
          response to glucose
          post-embryonic development
          embryonic pattern specification
          anterior/posterior pattern specification
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          regulation of transforming growth factor beta receptor signaling pathway
          signal transduction involved in regulation of gene expression
          insulin secretion
          lung development
          positive regulation of BMP signaling pathway
          pancreas development
          activin receptor signaling pathway
          organ growth
          intracellular signal transduction
          nodal signaling pathway
          cell fate commitment
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          paraxial mesoderm morphogenesis
          developmental growth
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of binding
          palate development
          pericardium development
          SMAD protein signal transduction
          response to cholesterol
          positive regulation of nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          transforming growth factor beta receptor binding
          protein binding
          transcription factor binding
          phosphatase binding
          transforming growth factor beta receptor, pathway-specific cytoplasmic mediator activity
          ubiquitin protein ligase binding
          activating transcription factor binding
          type I transforming growth factor beta receptor binding
          protein homodimerization activity
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          co-SMAD binding
          I-SMAD binding
          R-SMAD binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P210520 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 142118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868884 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 679192 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200444 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564550 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884865 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 332139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249149 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553519 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576664 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 935346 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 269342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189323 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 518813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 504557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 200106 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35490 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 491092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 481349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971746 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 762170 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55725 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 572899 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459783 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 530588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56183 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 59701 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 135853 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 133277 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 152419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166627 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132371 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35143 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P210520 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P210520 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P210520 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P210520 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700312754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58416 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61989 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5609 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10205 - -
          Agilent Rat cDNA Microarray (G4105A) 700312754 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10802744 - -
          Affymetrix Rat Gene 1.0 ST Array 10802754 - -
          Affymetrix Rat Gene 1.0 ST Array 10802751 - -
          Affymetrix Rat Gene 1.0 ST Array 10802747 - -
          Affymetrix Rat Gene 1.0 ST Array 10802752 - -
          Affymetrix Rat Gene 1.0 ST Array 10802746 - -
          Affymetrix Rat Gene 1.0 ST Array 10802745 - -
          Affymetrix Rat Gene 1.0 ST Array 10802753 - -
          Affymetrix Rat Gene 1.0 ST Array 10802748 - -
          Affymetrix Rat Gene 1.0 ST Array 10802749 - -
          Affymetrix Rat Gene 1.0 ST Array 10802743 - -
          Affymetrix Rat Gene 1.0 ST Array 10802750 - -
          Affymetrix Rat Gene 1.1 ST Array 10802741 - -
          Rat Genome 230 2.0 Array 1368214_at - -
          Rat Genome 230 2.0 Array 1368214_PM_at - -
          Affymetrix HT Rat Focus Array 1368214_PM_at - -
          Affymetrix HT Rat Focus Array 1368214_at - -
          Affymetrix Rat Clariom S Assay PSR1800006049.rn.1 TC1800000839.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800006052.rn.1 TC1800000839.rn.1 RTA TC
          Rat Expression Set 230 1368214_PM_at - -
          Rat Expression Set 230 1368214_at - -
          Affymetrix Rat Gene 1.0 ST Array 10802741 - -
          Rat Genome U34 Set AB017912_at - -
          Rat Genome U34 Set AB017912_g_at - -
          Rat Genome U34 Set rc_AI228675_at - -
          Rat Genome U34 Set rc_AI105449_at - -
          Rat Genome U34 Set AB017912_at - -
          Rat Genome U34 Set AB017912_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21597126 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360346 - -
          RatRef-12 Expression BeadChip ILMN_65079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline