Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570060_g1
          See other ACTA1 GE Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570060_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019212.2 NP_062085.1 5 - 6 870 71
          GenBank mRNA BC061974.1 - 5 - 6 870 5623909 71
          FQ214598.1 - 5 - 6 880 72
          FQ214600.1 - 5 - 6 72
          FQ214610.1 - 3 - 4 424 71
          FQ214623.1 - 5 - 6 878 71
          FQ214659.1 - 5 - 6 881 71
          FQ214676.1 - 5 - 6 880 71
          FQ214734.1 - 5 - 6 880 71
          FQ214743.1 - 5 - 6 879 71
          FQ214761.1 - 5 - 6 878 71
          FQ214786.1 - 5 - 6 882 71
          FQ214858.1 - 5 - 6 879 71
          FQ214883.1 - 5 - 6 878 71
          FQ214904.1 - 5 - 6 879 71
          FQ214948.1 - 5 - 6 879 71
          FQ214949.1 - 5 - 6 880 71
          FQ214969.1 - 5 - 6 879 71
          FQ214976.1 - 5 - 6 879 71
          FQ214987.1 - 5 - 6 879 71
          FQ214993.1 - 5 - 6 879 71
          FQ215012.1 - 5 - 6 880 71
          FQ215038.1 - 5 - 6 878 71
          FQ215058.1 - 5 - 6 880 71
          FQ215089.1 - 5 - 6 880 71
          FQ215136.1 - 5 - 6 879 71
          FQ215185.1 - 5 - 6 879 71
          FQ215204.1 - 5 - 6 877 71
          FQ215241.1 - 5 - 6 880 71
          FQ215249.1 - 5 - 6 880 71
          FQ215256.1 - 5 - 6 879 71
          FQ215282.1 - 5 - 6 879 71
          FQ215298.1 - 5 - 6 880 71
          FQ215324.1 - 5 - 6 879 71
          FQ215380.1 - 5 - 6 881 71
          FQ215396.1 - 5 - 6 879 71
          FQ215398.1 - 5 - 6 879 71
          FQ215431.1 - 5 - 6 880 71
          FQ215464.1 - 5 - 6 879 71
          FQ215465.1 - 5 - 6 879 71
          FQ215470.1 - 5 - 6 879 71
          FQ215488.1 - 5 - 6 879 71
          FQ215518.1 - 5 - 6 888 71
          FQ215554.1 - 5 - 6 879 71
          FQ215618.1 - 5 - 6 880 71
          FQ215628.1 - 5 - 6 881 72
          FQ215631.1 - 3 - 4 493 71
          FQ215659.1 - 5 - 6 878 71
          FQ215673.1 - 5 - 6 879 71
          FQ215690.1 - 5 - 6 880 71
          FQ215809.1 - 5 - 6 879 71
          FQ215813.1 - 5 - 6 879 71
          FQ215836.1 - 5 - 6 879 71
          FQ215839.1 - 5 - 6 879 71
          FQ215841.1 - 5 - 6 880 71
          FQ215852.1 - 5 - 6 877 71
          FQ215864.1 - 5 - 6 880 72
          FQ215866.1 - 5 - 6 880 71
          FQ215888.1 - 5 - 6 878 71
          FQ215896.1 - 5 - 6 879 71
          FQ215924.1 - 3 - 4 571 71
          FQ216035.1 - 5 - 6 879 71
          FQ216043.1 - 5 - 6 879 71
          FQ216050.1 - 5 - 6 879 71
          FQ216224.1 - 5 - 6 880 71
          FQ216276.1 - 5 - 6 879 71
          FQ216291.1 - 5 - 6 880 71
          FQ216293.1 - 5 - 6 880 71
          FQ216360.1 - 5 - 6 880 71
          FQ216403.1 - 5 - 6 879 71
          FQ216430.1 - 5 - 6 880 71
          FQ216509.1 - 5 - 6 877 71
          FQ216603.1 - 5 - 6 881 71
          FQ216606.1 - 5 - 6 879 71
          FQ216607.1 - 5 - 6 875 71
          FQ216620.1 - 5 - 6 880 71
          FQ216639.1 - 5 - 6 879 71
          FQ216675.1 - 5 - 6 879 71
          FQ216676.1 - 5 - 6 879 71
          FQ216703.1 - 5 - 6 879 71
          FQ216735.1 - 1 - 2 119 71
          FQ216739.1 - 5 - 6 879 71
          FQ216767.1 - 5 - 6 879 71
          FQ216777.1 - 5 - 6 879 71
          FQ216793.1 - 5 - 6 879 71
          FQ216799.1 - 5 - 6 877 71
          FQ216801.1 - 5 - 6 879 71
          FQ216819.1 - 5 - 6 877 71
          FQ216918.1 - 3 - 4 522 71
          FQ216999.1 - 1 - 2 78 71
          FQ217061.1 - 5 - 6 881 71
          FQ217091.1 - 5 - 6 879 71
          FQ217173.1 - 5 - 6 879 71
          FQ217188.1 - 5 - 6 879 71
          FQ217194.1 - 5 - 6 879 71
          FQ217280.1 - 5 - 6 999 71
          FQ217329.1 - 5 - 6 879 71
          FQ217347.1 - 5 - 6 70
          FQ217353.1 - 5 - 6 879 71
          FQ217367.1 - 5 - 6 71
          FQ217441.1 - 5 - 6 879 71
          FQ217443.1 - 5 - 6 881 71
          FQ217475.1 - 5 - 6 880 71
          FQ217525.1 - 5 - 6 879 71
          FQ217535.1 - 5 - 6 879 71
          FQ217572.1 - 5 - 6 879 71
          FQ217615.1 - 5 - 6 879 71
          FQ217629.1 - 5 - 6 880 72
          FQ217668.1 - 1 - 2 128 71
          FQ217893.1 - 5 - 6 880 71
          FQ217919.1 - 5 - 6 879 71
          FQ217937.1 - 5 - 6 879 71
          FQ218005.1 - 2 - 3 279 71
          FQ218019.1 - 5 - 6 880 71
          FQ223686.1 - 5 - 6 879 71
          FQ223702.1 - 5 - 6 880 71
          FQ223727.1 - 3 - 4 577 71
          FQ223778.1 - 3 - 4 393 71
          FQ223926.1 - 5 - 6 879 71
          FQ224082.1 - 5 - 6 879 71
          FQ224862.1 - 5 - 6 879 71
          FQ224886.1 - 5 - 6 878 71
          FQ224890.1 - 5 - 6 877 71
          FQ224936.1 - 5 - 6 880 71
          FQ224946.1 - 3 - 4 407 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001667.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acta1

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P255236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 473877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 898527 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255236 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P255236 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P255236 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P255236 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811828 - -
          Affymetrix Rat Gene 1.0 ST Array 10811831 - -
          Affymetrix Rat Gene 1.0 ST Array 10811827 - -
          Affymetrix Rat Gene 1.0 ST Array 10811826 - -
          Affymetrix Rat Gene 1.0 ST Array 10811830 - -
          Affymetrix Rat Gene 1.0 ST Array 10811825 - -
          Affymetrix Rat Gene 1.0 ST Array 10811829 - -
          Affymetrix Rat Gene 1.1 ST Array 10811824 - -
          Rat Genome 230 2.0 Array 1369928_at - -
          Rat Genome 230 2.0 Array 1385797_PM_at - -
          Rat Genome 230 2.0 Array 1385797_at - -
          Rat Genome 230 2.0 Array 1369928_PM_at - -
          Affymetrix HT Rat Focus Array 1369928_PM_at - -
          Affymetrix HT Rat Focus Array 1369928_at - -
          Affymetrix Rat Clariom S Assay PSR1900013467.rn.1 TC1900001667.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013468.rn.1 TC1900001667.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013469.rn.1 TC1900001667.rn.1 RTA TC
          Rat Expression Set 230 1369928_PM_at - -
          Rat Expression Set 230 1369928_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811824 - -
          Rat Genome U34 Set J00692_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21344519 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360149 - -
          RatRef-12 Expression BeadChip ILMN_48417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline