Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570707_m1
          See other CAR5A GE Assays ›
          Gene Symbol:
          Car5a
          Gene Name:
          carbonic anhydrase 5a, mitochondrial
          Gene Aliases:
          Ca5, Ca5a
          Chromosome Location:
          Chr.19: 54731838 - 54761695 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570707_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Car5a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq XM_006255782.2 XP_006255844.1 3 - 4 565 100
          GenBank mRNA BC088147.1 - 3 - 4 507 7323596 100
          U12268.1 - 3 - 4 470 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900001619.rn.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          one-carbon metabolic process
          carbonate dehydratase activity
          zinc ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P671800 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352830 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 339503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43770 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288228 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 301299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832341 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141825 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 132010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 623809 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 155071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150187 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9319 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671800 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P671800 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P671800 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P671800 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104783 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51760 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48306 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66677 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10811548 - -
          Affymetrix Rat Gene 1.0 ST Array 10811550 - -
          Affymetrix Rat Gene 1.0 ST Array 10811546 - -
          Affymetrix Rat Gene 1.0 ST Array 10811549 - -
          Affymetrix Rat Gene 1.0 ST Array 10811545 - -
          Affymetrix Rat Gene 1.0 ST Array 10811547 - -
          Affymetrix Rat Gene 1.0 ST Array 10811544 - -
          Affymetrix Rat Gene 1.0 ST Array 10811551 - -
          Affymetrix Rat Gene 1.1 ST Array 10811543 - -
          Rat Genome 230 2.0 Array 1387609_at - -
          Rat Genome 230 2.0 Array 1387609_PM_at - -
          Affymetrix HT Rat Focus Array 1387609_at - -
          Affymetrix HT Rat Focus Array 1387609_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900013035.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013034.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013032.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013033.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013036.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013030.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013029.rn.1 TC1900001619.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900013031.rn.1 TC1900001619.rn.1 RTA TC
          Rat Expression Set 230 1387609_PM_at - -
          Rat Expression Set 230 1387609_at - -
          Affymetrix Rat Gene 1.0 ST Array 10811543 - -
          Rat Genome U34 Set U12268_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21434587 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362319 - -
          RatRef-12 Expression BeadChip ILMN_58560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline