Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00570740_m1
          See other CHRND GE Assays ›
          Gene Symbol:
          Chrnd
          Gene Name:
          cholinergic receptor, nicotinic, delta (muscle)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 94286550 - 94294968 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00570740_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrnd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019298.1 NP_062171.1 6 - 7 616 94
          GenBank mRNA X74835.1 - 6 - 7 616 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900002708.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrnd

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal muscle contraction
          cation transport
          ion transmembrane transport
          regulation of membrane potential
          skeletal muscle tissue growth
          musculoskeletal movement
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          ligand-gated ion channel activity
          acetylcholine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P116652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678558 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416508 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 53080 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P116652 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P116652 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P116652 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P116652 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 12246 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924978 - -
          Affymetrix Rat Gene 1.0 ST Array 10924977 - -
          Affymetrix Rat Gene 1.0 ST Array 10924975 - -
          Affymetrix Rat Gene 1.0 ST Array 10924980 - -
          Affymetrix Rat Gene 1.0 ST Array 10924979 - -
          Affymetrix Rat Gene 1.0 ST Array 10924984 - -
          Affymetrix Rat Gene 1.0 ST Array 10924983 - -
          Affymetrix Rat Gene 1.0 ST Array 10924988 - -
          Affymetrix Rat Gene 1.0 ST Array 10924982 - -
          Affymetrix Rat Gene 1.0 ST Array 10924985 - -
          Affymetrix Rat Gene 1.0 ST Array 10924986 - -
          Affymetrix Rat Gene 1.0 ST Array 10924987 - -
          Affymetrix Rat Gene 1.0 ST Array 10924981 - -
          Affymetrix Rat Gene 1.1 ST Array 10924974 - -
          Rat Genome 230 2.0 Array 1368734_at - -
          Rat Genome 230 2.0 Array 1368734_PM_at - -
          Affymetrix HT Rat Focus Array 1368734_at - -
          Affymetrix HT Rat Focus Array 1368734_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900022239.rn.1 TC0900002708.rn.1 RTA TC
          Rat Expression Set 230 1368734_at - -
          Rat Expression Set 230 1368734_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924974 - -
          Rat Genome U34 Set X74835cds_at - -
          Rat Genome U34 Set X74835cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21794588 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366954 - -
          RatRef-12 Expression BeadChip ILMN_62009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline