Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00571492_m1
          See other CYP4F1 GE Assays ›
          Gene Symbol:
          Cyp4f1
          Gene Name:
          cytochrome P450, family 4, subfamily f, polypeptide 1
          Gene Aliases:
          Cyp4f14, Cyp4f2
          Chromosome Location:
          Chr.7: 15179472 - 15191111 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00571492_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4f1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019623.2 NP_062569.2 2 - 3 476 80
          XM_008765180.1 XP_008763402.1 3 - 4 427 80
          XM_008765181.1 XP_008763403.1 3 - 4 370 80
          GenBank mRNA BC081808.1 - 2 - 3 476 7109036 80
          M94548.1 - 2 - 3 369 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002254.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection P450 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp4f1

          Panther Classification:

          Molecular Function -

          oxygenase

          Gene Ontology Categories:

          Function(s) Process(es)

          icosanoid metabolic process
          response to toxic substance
          arachidonic acid metabolic process
          oxidation-reduction process
          monooxygenase activity
          iron ion binding
          arachidonic acid monooxygenase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          arachidonic acid binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 333713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125966 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144457 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401973 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120947 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12051 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12051 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12051 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12051 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700930836 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39972 - -
          Agilent Rat cDNA Microarray (G4105A) 700930836 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10901058 - -
          Affymetrix Rat Gene 1.0 ST Array 10901063 - -
          Affymetrix Rat Gene 1.0 ST Array 10901062 - -
          Affymetrix Rat Gene 1.0 ST Array 10901057 - -
          Affymetrix Rat Gene 1.0 ST Array 10901064 - -
          Affymetrix Rat Gene 1.0 ST Array 10901060 - -
          Affymetrix Rat Gene 1.0 ST Array 10901067 - -
          Affymetrix Rat Gene 1.0 ST Array 10901061 - -
          Affymetrix Rat Gene 1.0 ST Array 10901065 - -
          Affymetrix Rat Gene 1.0 ST Array 10901059 - -
          Affymetrix Rat Gene 1.0 ST Array 10901068 - -
          Affymetrix Rat Gene 1.0 ST Array 10901056 - -
          Affymetrix Rat Gene 1.0 ST Array 10901066 - -
          Affymetrix Rat Gene 1.1 ST Array 10901055 - -
          Rat Genome 230 2.0 Array 1368467_at - -
          Rat Genome 230 2.0 Array 1368467_PM_at - -
          Affymetrix HT Rat Focus Array 1368467_at - -
          Affymetrix HT Rat Focus Array 1368467_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700018151.rn.1 TC0700002254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018153.rn.1 TC0700002254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018154.rn.1 TC0700002254.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700018155.rn.1 TC0700002254.rn.1 RTA TC
          Rat Expression Set 230 1368467_at - -
          Rat Expression Set 230 1368467_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10901055 - -
          Rat Genome U34 Set M94548_at - -
          Rat Genome U34 Set M94548_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21977747 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372735 - -
          RatRef-12 Expression BeadChip ILMN_64548 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline