Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572394_m1
          See other ADCY10 GE Assays ›
          Gene Symbol:
          Adcy10
          Gene Name:
          adenylate cyclase 10 (soluble)
          Gene Aliases:
          Sac
          Chromosome Location:
          Chr.13: 83701970 - 83787010 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572394_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adcy10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021684.1 NP_067716.1 20 - 21 2744 83
          XM_008769677.1 XP_008767899.1 19 - 20 4791 83
          XM_008769678.1 XP_008767900.1 19 - 20 4788 83
          XM_008769679.1 XP_008767901.1 19 - 20 2517 83
          XM_008769680.1 XP_008767902.1 17 - 18 4605 83
          GenBank mRNA AF081941.1 - 20 - 21 2744 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300000668.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adcy10

          Gene Ontology Categories:

          Function(s) Process(es)

          cAMP biosynthetic process
          spermatogenesis
          intracellular signal transduction
          positive regulation of apoptotic process
          magnesium ion binding
          adenylate cyclase activity
          ATP binding
          manganese ion binding
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P460021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37468 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231380 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625391 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 350963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658076 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360761 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551662 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947570 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474980 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734226 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 145991 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 812342 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 180989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 718387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856670 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913548 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 329911 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 316703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325673 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 815677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 415446 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532595 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306792 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 779652 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 236419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 607603 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114067 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168034 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74599 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 104790 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 76638 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154769 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93960 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91661 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134775 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 155545 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51063 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96877 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P460021 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P460021 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P460021 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P460021 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56408 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59454 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86743 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58969 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93436 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77190 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 41403 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90576 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10765323 - -
          Affymetrix Rat Gene 1.0 ST Array 10765322 - -
          Affymetrix Rat Gene 1.0 ST Array 10765309 - -
          Affymetrix Rat Gene 1.0 ST Array 10765321 - -
          Affymetrix Rat Gene 1.0 ST Array 10765308 - -
          Affymetrix Rat Gene 1.0 ST Array 10765320 - -
          Affymetrix Rat Gene 1.0 ST Array 10765333 - -
          Affymetrix Rat Gene 1.0 ST Array 10765310 - -
          Affymetrix Rat Gene 1.0 ST Array 10765325 - -
          Affymetrix Rat Gene 1.0 ST Array 10765332 - -
          Affymetrix Rat Gene 1.0 ST Array 10765304 - -
          Affymetrix Rat Gene 1.0 ST Array 10765306 - -
          Affymetrix Rat Gene 1.0 ST Array 10765301 - -
          Affymetrix Rat Gene 1.0 ST Array 10765311 - -
          Affymetrix Rat Gene 1.0 ST Array 10765307 - -
          Affymetrix Rat Gene 1.0 ST Array 10765316 - -
          Affymetrix Rat Gene 1.0 ST Array 10765328 - -
          Affymetrix Rat Gene 1.0 ST Array 10765314 - -
          Affymetrix Rat Gene 1.0 ST Array 10765302 - -
          Affymetrix Rat Gene 1.0 ST Array 10765305 - -
          Affymetrix Rat Gene 1.0 ST Array 10765334 - -
          Affymetrix Rat Gene 1.0 ST Array 10765326 - -
          Affymetrix Rat Gene 1.0 ST Array 10765331 - -
          Affymetrix Rat Gene 1.0 ST Array 10765329 - -
          Affymetrix Rat Gene 1.0 ST Array 10765299 - -
          Affymetrix Rat Gene 1.0 ST Array 10765317 - -
          Affymetrix Rat Gene 1.0 ST Array 10765303 - -
          Affymetrix Rat Gene 1.0 ST Array 10765315 - -
          Affymetrix Rat Gene 1.0 ST Array 10765330 - -
          Affymetrix Rat Gene 1.0 ST Array 10765327 - -
          Affymetrix Rat Gene 1.0 ST Array 10765318 - -
          Affymetrix Rat Gene 1.0 ST Array 10765313 - -
          Affymetrix Rat Gene 1.0 ST Array 10765312 - -
          Affymetrix Rat Gene 1.0 ST Array 10765300 - -
          Affymetrix Rat Gene 1.0 ST Array 10765319 - -
          Affymetrix Rat Gene 1.0 ST Array 10765324 - -
          Affymetrix Rat Gene 1.1 ST Array 10765298 - -
          Rat Genome 230 2.0 Array 1369337_PM_at - -
          Rat Genome 230 2.0 Array 1369337_at - -
          Affymetrix HT Rat Focus Array 1369337_PM_at - -
          Affymetrix HT Rat Focus Array 1369337_at - -
          Affymetrix Rat Clariom S Assay PSR1300004715.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004712.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004713.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004714.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004716.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004717.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004710.rn.1 TC1300000668.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1300004718.rn.1 TC1300000668.rn.1 RTA TC
          Rat Expression Set 230 1369337_at - -
          Rat Expression Set 230 1369337_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10765298 - -
          Rat Genome U34 Set AF081941_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21594326 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline