Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572463_m1
          See other RAPGEF3 GE Assays ›
          Gene Symbol:
          Rapgef3
          Gene Name:
          Rap guanine nucleotide exchange factor (GEF) 3
          Gene Aliases:
          Epac
          Chromosome Location:
          Chr.7: 139232263 - 139254883 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572463_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rapgef3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021690.1 NP_067722.1 15 - 16 1672 65
          XM_006242352.2 XP_006242414.1 16 - 17 1932 65
          XM_008765778.1 XP_008764000.1 16 - 17 2221 65
          XM_008765779.1 XP_008764001.1 16 - 17 1839 65
          XM_008765780.1 XP_008764002.1 15 - 16 2179 65
          GenBank mRNA BC107667.1 - 15 - 16 1625 7936411 65
          U78167.1 - 15 - 16 1672 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003822.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rapgef3

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          Rap protein signal transduction
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of syncytium formation by plasma membrane fusion
          positive regulation of GTPase activity
          positive regulation of angiogenesis
          regulation of protein kinase activity
          positive regulation of protein export from nucleus
          positive regulation of stress fiber assembly
          positive regulation of syncytium formation by plasma membrane fusion
          establishment of endothelial barrier
          cellular response to cAMP
          positive regulation of protein acetylation
          regulation of actin cytoskeleton reorganization
          regulation of histone H3-K9 acetylation
          guanyl-nucleotide exchange factor activity
          cAMP-dependent protein kinase regulator activity
          Rap guanyl-nucleotide exchange factor activity
          cAMP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 115155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445545 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630979 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 555696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 913559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859172 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897612 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 680681 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953015 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 85138 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14357 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15424 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15424 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15424 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P15424 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103975 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 51979 - -
          Agilent Rat cDNA Microarray (G4105A) 700777145 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10906695 - -
          Affymetrix Rat Gene 1.0 ST Array 10906705 - -
          Affymetrix Rat Gene 1.0 ST Array 10906697 - -
          Affymetrix Rat Gene 1.0 ST Array 10906696 - -
          Affymetrix Rat Gene 1.0 ST Array 10906707 - -
          Affymetrix Rat Gene 1.0 ST Array 10906694 - -
          Affymetrix Rat Gene 1.0 ST Array 10906688 - -
          Affymetrix Rat Gene 1.0 ST Array 10906702 - -
          Affymetrix Rat Gene 1.0 ST Array 10906684 - -
          Affymetrix Rat Gene 1.0 ST Array 10906706 - -
          Affymetrix Rat Gene 1.0 ST Array 10906686 - -
          Affymetrix Rat Gene 1.0 ST Array 10906687 - -
          Affymetrix Rat Gene 1.0 ST Array 10906692 - -
          Affymetrix Rat Gene 1.0 ST Array 10906690 - -
          Affymetrix Rat Gene 1.0 ST Array 10906680 - -
          Affymetrix Rat Gene 1.0 ST Array 10906685 - -
          Affymetrix Rat Gene 1.0 ST Array 10906691 - -
          Affymetrix Rat Gene 1.0 ST Array 10906693 - -
          Affymetrix Rat Gene 1.0 ST Array 10906681 - -
          Affymetrix Rat Gene 1.0 ST Array 10906704 - -
          Affymetrix Rat Gene 1.0 ST Array 10906700 - -
          Affymetrix Rat Gene 1.0 ST Array 10906683 - -
          Affymetrix Rat Gene 1.0 ST Array 10906689 - -
          Affymetrix Rat Gene 1.0 ST Array 10906703 - -
          Affymetrix Rat Gene 1.0 ST Array 10906699 - -
          Affymetrix Rat Gene 1.0 ST Array 10898950 - -
          Affymetrix Rat Gene 1.0 ST Array 10906698 - -
          Affymetrix Rat Gene 1.0 ST Array 10906682 - -
          Affymetrix Rat Gene 1.1 ST Array 10906679 - -
          Rat Genome 230 2.0 Array 1368660_PM_at - -
          Rat Genome 230 2.0 Array 1368660_at - -
          Affymetrix HT Rat Focus Array 1368660_at - -
          Affymetrix HT Rat Focus Array 1368660_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0700030505.rn.1 TC0700003822.rn.1 RTA TC
          Rat Expression Set 230 1368660_at - -
          Rat Expression Set 230 1368660_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10906679 - -
          Rat Genome U34 Set U78167_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21913955 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357364 - -
          RatRef-12 Expression BeadChip ILMN_59861 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline