Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00572643_m1
          See other CEP78 GE Assays ›
          Gene Symbol:
          Cep78
          Gene Name:
          centrosomal protein 78
          Gene Aliases:
          Ip63
          Chromosome Location:
          Chr.1: 233185815 - 233214873 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00572643_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cep78
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021741.2 NP_068509.2 5 - 6 794 78
          XM_006231136.2 XP_006231198.1 5 - 6 909 78
          GenBank mRNA FQ222521.1 - 3 - 4 659 78
          X99330.1 - 5 - 6 582 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100007350.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cep78

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P580618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 239147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943184 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 918129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19883 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 510171 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622269 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917898 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 883325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625423 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167226 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148369 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 8437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70501 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P580618 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P580618 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P580618 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P580618 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700530863 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76656 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47325 - -
          Agilent Rat cDNA Microarray (G4105A) 700530863 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10729114 - -
          Affymetrix Rat Gene 1.0 ST Array 10729119 - -
          Affymetrix Rat Gene 1.0 ST Array 10729113 - -
          Affymetrix Rat Gene 1.0 ST Array 10729115 - -
          Affymetrix Rat Gene 1.0 ST Array 10729108 - -
          Affymetrix Rat Gene 1.0 ST Array 10729110 - -
          Affymetrix Rat Gene 1.0 ST Array 10729103 - -
          Affymetrix Rat Gene 1.0 ST Array 10729104 - -
          Affymetrix Rat Gene 1.0 ST Array 10729117 - -
          Affymetrix Rat Gene 1.0 ST Array 10729107 - -
          Affymetrix Rat Gene 1.0 ST Array 10729106 - -
          Affymetrix Rat Gene 1.0 ST Array 10729111 - -
          Affymetrix Rat Gene 1.0 ST Array 10729112 - -
          Affymetrix Rat Gene 1.0 ST Array 10729116 - -
          Affymetrix Rat Gene 1.0 ST Array 10729105 - -
          Affymetrix Rat Gene 1.0 ST Array 10729109 - -
          Affymetrix Rat Gene 1.1 ST Array 10729102 - -
          Rat Genome 230 2.0 Array 1369433_PM_at - -
          Rat Genome 230 2.0 Array 1369433_at - -
          Affymetrix HT Rat Focus Array 1369433_PM_at - -
          Affymetrix HT Rat Focus Array 1369433_at - -
          Affymetrix Rat Clariom S Assay PSR0100059381.rn.1 TC0100007350.rn.1 RTA TC
          Rat Expression Set 230 1369433_PM_at - -
          Rat Expression Set 230 1369433_at - -
          Affymetrix Rat Gene 1.0 ST Array 10729102 - -
          Rat Genome U34 Set X99330cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21274703 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline