Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573159_m1
          See other MATK GE Assays ›
          Gene Symbol:
          Matk
          Gene Name:
          megakaryocyte-associated tyrosine kinase
          Gene Aliases:
          Batk
          Chromosome Location:
          Chr.7: 11325246 - 11334300 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573159_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Matk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_021859.2 NP_068631.1 7 - 8 724 88
          XM_006240989.2 XP_006241051.1 7 - 8 1090 88
          XM_006240990.2 XP_006241052.1 7 - 8 750 88
          XM_008765128.1 XP_008763350.1 7 - 8 1054 88
          GenBank mRNA BC087726.1 - 7 - 8 724 7320205 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700002116.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Matk

          Gene Ontology Categories:

          Function(s) Process(es)

          morphogenesis of an epithelium
          transmembrane receptor protein tyrosine kinase signaling pathway
          cell migration
          cell differentiation
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          innate immune response
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P466321 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 754720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 349098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345747 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P466321 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P466321 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P466321 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P466321 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32577 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10900276 - -
          Affymetrix Rat Gene 1.0 ST Array 10900267 - -
          Affymetrix Rat Gene 1.0 ST Array 10900278 - -
          Affymetrix Rat Gene 1.0 ST Array 10900271 - -
          Affymetrix Rat Gene 1.0 ST Array 10900264 - -
          Affymetrix Rat Gene 1.0 ST Array 10900272 - -
          Affymetrix Rat Gene 1.0 ST Array 10900265 - -
          Affymetrix Rat Gene 1.0 ST Array 10900273 - -
          Affymetrix Rat Gene 1.0 ST Array 10900274 - -
          Affymetrix Rat Gene 1.0 ST Array 10900270 - -
          Affymetrix Rat Gene 1.0 ST Array 10900275 - -
          Affymetrix Rat Gene 1.0 ST Array 10900269 - -
          Affymetrix Rat Gene 1.0 ST Array 10900266 - -
          Affymetrix Rat Gene 1.1 ST Array 10900263 - -
          Rat Genome 230 2.0 Array 1387834_PM_at - -
          Rat Genome 230 2.0 Array 1387834_at - -
          Affymetrix HT Rat Focus Array 1387834_PM_at - -
          Affymetrix HT Rat Focus Array 1387834_at - -
          Affymetrix Rat Clariom S Assay PSR0700016483.rn.1 TC0700002116.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700016482.rn.1 TC0700002116.rn.1 RTA TC
          Rat Expression Set 230 1387834_PM_at - -
          Rat Expression Set 230 1387834_at - -
          Affymetrix Rat Gene 1.0 ST Array 10900263 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21394104 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372033 - -
          RatRef-12 Expression BeadChip ILMN_62543 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline