Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573318_m1
          See other AQP6 GE Assays ›
          Gene Symbol:
          Aqp6
          Gene Name:
          aquaporin 6, kidney specific
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 141258585 - 141261518 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573318_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aqp6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022181.1 NP_071517.1 3 - 4 773 61
          GenBank mRNA AF083879.1 - 3 - 4 773 61
          L28113.1 - 3 - 4 800 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001819.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aqp6

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          water transport
          cellular water homeostasis
          nitrate transport
          glycerol transport
          ion transmembrane transport
          odontogenesis
          anion transmembrane transport
          anion channel activity
          nitrate transmembrane transporter activity
          water channel activity
          glycerol channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P122362 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307221 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98900 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122362 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P122362 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P122362 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P558451 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 103422 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75340 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19012 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75790 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1626 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 37955 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 80839 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10899144 - -
          Affymetrix Rat Gene 1.0 ST Array 10899140 - -
          Affymetrix Rat Gene 1.0 ST Array 10899141 - -
          Affymetrix Rat Gene 1.0 ST Array 10899142 - -
          Affymetrix Rat Gene 1.0 ST Array 10899143 - -
          Affymetrix Rat Gene 1.1 ST Array 10899139 - -
          Rat Genome 230 2.0 Array 1369298_PM_at - -
          Rat Genome 230 2.0 Array 1369298_at - -
          Affymetrix HT Rat Focus Array 1369298_PM_at - -
          Affymetrix HT Rat Focus Array 1369298_at - -
          Affymetrix Rat Clariom S Assay PSR0700014032.rn.1 TC0700001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014029.rn.1 TC0700001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014030.rn.1 TC0700001819.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700014031.rn.1 TC0700001819.rn.1 RTA TC
          Rat Expression Set 230 1369298_PM_at - -
          Rat Expression Set 230 1369298_at - -
          Affymetrix Rat Gene 1.0 ST Array 10899139 - -
          Rat Genome U34 Set L28113_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22126943 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1354810 - -
          RatRef-12 Expression BeadChip ILMN_69913 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline