Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00573424_m1
          See other ACAN GE Assays ›
          Gene Symbol:
          Acan
          Gene Name:
          aggrecan
          Gene Aliases:
          Agc, Agc1
          Chromosome Location:
          Chr.1: 140762758 - 140824441 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00573424_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acan
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022190.1 NP_071526.1 2 - 3 454 74
          GenBank mRNA J03485.1 - 2 - 3 454 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100001823.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acan

          Panther Classification:

          Molecular Function -

          extracellular matrix glycoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          cartilage condensation
          ossification
          chondrocyte development
          cell adhesion
          central nervous system development
          response to radiation
          response to mechanical stimulus
          response to gravity
          response to glucose
          multicellular organism aging
          response to acidic pH
          response to organic cyclic compound
          spinal cord development
          proteoglycan biosynthetic process
          collagen fibril organization
          negative regulation of cell migration
          response to drug
          chondroblast differentiation
          cellular response to growth factor stimulus
          calcium ion binding
          hyaluronic acid binding
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908452 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450005 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 672955 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 50382 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 320530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 117189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 306185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 56618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613051 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 470910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 870767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 247744 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871328 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46046 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 368432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843984 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189721 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 464074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 941012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123827 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266397 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 622101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337817 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908837 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 800091 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784646 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 164233 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520846 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125642 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472733 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489904 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 774512 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 147680 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55941 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164072 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75058 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29949 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 52233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81957 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 9218 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12154 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12154 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12154 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12154 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 43204 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 49678 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 74604 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10708044 - -
          Affymetrix Rat Gene 1.0 ST Array 10708036 - -
          Affymetrix Rat Gene 1.0 ST Array 10708028 - -
          Affymetrix Rat Gene 1.0 ST Array 10708043 - -
          Affymetrix Rat Gene 1.0 ST Array 10708040 - -
          Affymetrix Rat Gene 1.0 ST Array 10708026 - -
          Affymetrix Rat Gene 1.0 ST Array 10708025 - -
          Affymetrix Rat Gene 1.0 ST Array 10708035 - -
          Affymetrix Rat Gene 1.0 ST Array 10708049 - -
          Affymetrix Rat Gene 1.0 ST Array 10708038 - -
          Affymetrix Rat Gene 1.0 ST Array 10708050 - -
          Affymetrix Rat Gene 1.0 ST Array 10708027 - -
          Affymetrix Rat Gene 1.0 ST Array 10708030 - -
          Affymetrix Rat Gene 1.0 ST Array 10708022 - -
          Affymetrix Rat Gene 1.0 ST Array 10708048 - -
          Affymetrix Rat Gene 1.0 ST Array 10708046 - -
          Affymetrix Rat Gene 1.0 ST Array 10708042 - -
          Affymetrix Rat Gene 1.0 ST Array 10708031 - -
          Affymetrix Rat Gene 1.0 ST Array 10708023 - -
          Affymetrix Rat Gene 1.0 ST Array 10708034 - -
          Affymetrix Rat Gene 1.0 ST Array 10708039 - -
          Affymetrix Rat Gene 1.0 ST Array 10708041 - -
          Affymetrix Rat Gene 1.0 ST Array 10708047 - -
          Affymetrix Rat Gene 1.0 ST Array 10708029 - -
          Affymetrix Rat Gene 1.0 ST Array 10708024 - -
          Affymetrix Rat Gene 1.0 ST Array 10708032 - -
          Affymetrix Rat Gene 1.0 ST Array 10708033 - -
          Affymetrix Rat Gene 1.0 ST Array 10708045 - -
          Affymetrix Rat Gene 1.0 ST Array 10708052 - -
          Affymetrix Rat Gene 1.1 ST Array 10708021 - -
          Rat Genome 230 2.0 Array 1387355_at - -
          Rat Genome 230 2.0 Array 1368836_a_at - -
          Rat Genome 230 2.0 Array 1387355_PM_at - -
          Rat Genome 230 2.0 Array 1368836_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387355_at - -
          Affymetrix HT Rat Focus Array 1368836_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387355_PM_at - -
          Affymetrix HT Rat Focus Array 1368836_a_at - -
          Affymetrix Rat Clariom S Assay PSR0100014286.rn.1 TC0100001823.rn.1 RTA TC
          Rat Expression Set 230 1387355_at - -
          Rat Expression Set 230 1387355_PM_at - -
          Rat Expression Set 230 1368836_PM_a_at - -
          Rat Expression Set 230 1368836_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10708021 - -
          Rat Genome U34 Set rc_AA899552_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21332021 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1367163 - -
          RatRef-12 Expression BeadChip ILMN_57514 - -

          Back To Top
           

          Citations (1)

          Regulation of Sox9 activity by crosstalk with nuclear factor-kappaB and retinoic acid receptors.
          Authors
          Rockel JS, Kudirka JC, Guzi AJ, Bernier SM
          Journal
          Arthritis Res Ther - Page No R3 (Tue Jan 01 00:00:00 EST 2008)
          Volume (Year)
          10 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline