Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00574184_m1
          See other ELTD1 GE Assays ›
          Gene Symbol:
          Eltd1
          Gene Name:
          EGF, latrophilin and seven transmembrane domain containing 1
          Gene Aliases:
          Etl
          Chromosome Location:
          Chr.2: 256609787 - 256724642 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00574184_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eltd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022294.1 NP_071630.1 8 - 9 1260 99
          GenBank mRNA AF192401.1 - 8 - 9 1260 99
          AF192402.1 - 8 - 9 1260 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200002399.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 G-protein coupled receptor Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eltd1

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          G-protein coupled receptor activity
          calcium ion binding
          protein dimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P118447 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 171790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657753 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40356 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 878509 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 221552 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539285 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86538 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57526 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662742 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 90658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462993 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102379 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 243745 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 283243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 255934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475469 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 172080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209623 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446627 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 968788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 605420 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889310 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 226058 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 556943 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 35256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794784 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 286193 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 394813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 225723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882339 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325525 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 825197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 89719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 694167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 899831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360647 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 183788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143427 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 223074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 62607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33501 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150515 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93334 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130165 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125777 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137135 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28296 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47643 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 27466 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 126967 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172562 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118447 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12205 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P118447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P118447 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12205 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12205 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700522643 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28754 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 54225 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 20485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4028 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68822 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67032 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46388 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26688 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90374 - -
          Agilent Rat cDNA Microarray (G4105A) 700522643 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10819784 - -
          Affymetrix Rat Gene 1.0 ST Array 10819780 - -
          Affymetrix Rat Gene 1.0 ST Array 10819782 - -
          Affymetrix Rat Gene 1.0 ST Array 10819773 - -
          Affymetrix Rat Gene 1.0 ST Array 10819778 - -
          Affymetrix Rat Gene 1.0 ST Array 10819772 - -
          Affymetrix Rat Gene 1.0 ST Array 10819779 - -
          Affymetrix Rat Gene 1.0 ST Array 10819785 - -
          Affymetrix Rat Gene 1.0 ST Array 10819777 - -
          Affymetrix Rat Gene 1.0 ST Array 10819774 - -
          Affymetrix Rat Gene 1.0 ST Array 10819771 - -
          Affymetrix Rat Gene 1.0 ST Array 10819786 - -
          Affymetrix Rat Gene 1.0 ST Array 10819781 - -
          Affymetrix Rat Gene 1.0 ST Array 10819776 - -
          Affymetrix Rat Gene 1.0 ST Array 10819775 - -
          Affymetrix Rat Gene 1.0 ST Array 10819783 - -
          Affymetrix Rat Gene 1.1 ST Array 10819770 - -
          Rat Genome 230 2.0 Array 1387547_a_at - -
          Rat Genome 230 2.0 Array 1387547_PM_a_at - -
          Affymetrix HT Rat Focus Array 1387547_a_at - -
          Affymetrix HT Rat Focus Array 1387547_PM_a_at - -
          Affymetrix Rat Clariom S Assay PSR0200016836.rn.1 TC0200002399.rn.1 RTA TC
          Rat Expression Set 230 1387547_a_at - -
          Rat Expression Set 230 1387547_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10819770 - -
          Rat Genome U34 Set rc_AI237194_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21801833 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1369759 - -
          RatRef-12 Expression BeadChip ILMN_66456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline