Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575329_m1
          See other GRM4 GE Assays ›
          Gene Symbol:
          Grm4
          Gene Name:
          glutamate receptor, metabotropic 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 6745682 - 6791521 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575329_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022666.1 NP_073157.1 3546 69
          GenBank mRNA M90518.1 - 3546 69
          M92077.1 - 2761 69

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grm4

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          adenylate cyclase-inhibiting G-protein coupled glutamate receptor signaling pathway
          chemical synaptic transmission
          learning
          negative regulation of cell proliferation
          gamma-aminobutyric acid secretion
          neuron projection development
          synaptic transmission, glutamatergic
          negative regulation of apoptotic process
          positive regulation of MAPK cascade
          regulation of neuron apoptotic process
          negative regulation of synaptic transmission
          synaptic transmission, GABAergic
          regulation of synaptic transmission, glutamatergic
          group III metabotropic glutamate receptor activity
          G-protein coupled receptor activity
          calmodulin binding
          glutamate receptor activity
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P15406 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P163070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655741 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 857863 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 818599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509432 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 675693 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 768850 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657566 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 596279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 706003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233110 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 662159 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 715732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449958 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 509930 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126038 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 143663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83702 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115406 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 24628 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 158544 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15406 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163070 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P15406 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P163070 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P15406 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P163070 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P395757 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701921518 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 21466 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36685 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 7104 - -
          Agilent Rat cDNA Microarray (G4105A) 701921518 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10831850 - -
          Affymetrix Rat Gene 1.0 ST Array 10831854 - -
          Affymetrix Rat Gene 1.0 ST Array 10831852 - -
          Affymetrix Rat Gene 1.0 ST Array 10831856 - -
          Affymetrix Rat Gene 1.0 ST Array 10831858 - -
          Affymetrix Rat Gene 1.0 ST Array 10831857 - -
          Affymetrix Rat Gene 1.0 ST Array 10831855 - -
          Affymetrix Rat Gene 1.0 ST Array 10831853 - -
          Affymetrix Rat Gene 1.0 ST Array 10831851 - -
          Affymetrix Rat Gene 1.1 ST Array 10831849 - -
          Rat Genome 230 2.0 Array 1368690_PM_a_at - -
          Rat Genome 230 2.0 Array 1368690_a_at - -
          Affymetrix HT Rat Focus Array 1368690_a_at - -
          Affymetrix HT Rat Focus Array 1368690_PM_a_at - -
          Rat Expression Set 230 1368690_PM_a_at - -
          Rat Expression Set 230 1368690_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10831849 - -
          Rat Genome U34 Set M90518_at - -
          Rat Genome U34 Set M90518_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21895719 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349372 - -
          RatRef-12 Expression BeadChip ILMN_60401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline