Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00575768_m1
          See other B4GALNT1 GE Assays ›
          Gene Symbol:
          B4galnt1
          Gene Name:
          beta-1,4-N-acetyl-galactosaminyl transferase 1
          Gene Aliases:
          GalNAc-T, Galgt1
          Chromosome Location:
          Chr.7: 70449728 - 70459559 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00575768_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B4galnt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022860.2 NP_074051.1 6 - 7 857 67
          XM_006241497.2 XP_006241559.1 6 - 7 1384 67
          XM_008765419.1 XP_008763641.1 6 - 7 3213 67
          XM_008765420.1 XP_008763642.1 5 - 6 1534 67
          GenBank mRNA BC081799.1 - 6 - 7 857 7133837 67
          D17809.1 - 6 - 7 746 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000888.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene B4galnt1

          Gene Ontology Categories:

          Function(s) Process(es)

          ganglioside biosynthetic process
          protein glycosylation
          glycosphingolipid metabolic process
          spermatogenesis
          lipid storage
          lipid glycosylation
          (N-acetylneuraminyl)-galactosylglucosylceramide N-acetylgalactosaminyltransferase activity
          acetylgalactosaminyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P12325 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532221 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46342 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12325 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P12325 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P12325 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P12325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53042 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895836 - -
          Affymetrix Rat Gene 1.0 ST Array 10895843 - -
          Affymetrix Rat Gene 1.0 ST Array 10895837 - -
          Affymetrix Rat Gene 1.0 ST Array 10895842 - -
          Affymetrix Rat Gene 1.0 ST Array 10895841 - -
          Affymetrix Rat Gene 1.0 ST Array 10895833 - -
          Affymetrix Rat Gene 1.0 ST Array 10895835 - -
          Affymetrix Rat Gene 1.0 ST Array 10895834 - -
          Affymetrix Rat Gene 1.0 ST Array 10895839 - -
          Affymetrix Rat Gene 1.0 ST Array 10895832 - -
          Affymetrix Rat Gene 1.0 ST Array 10895840 - -
          Affymetrix Rat Gene 1.1 ST Array 10895830 - -
          Rat Genome 230 2.0 Array 1398272_at - -
          Rat Genome 230 2.0 Array 1398272_PM_at - -
          Affymetrix HT Rat Focus Array 1398272_PM_at - -
          Affymetrix HT Rat Focus Array 1398272_at - -
          Affymetrix Rat Clariom S Assay PSR0700006657.rn.1 TC0700000888.rn.1 RTA TC
          Rat Expression Set 230 1398272_at - -
          Rat Expression Set 230 1398272_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895830 - -
          Rat Genome U34 Set D17809_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21679082 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373152 - -
          RatRef-12 Expression BeadChip ILMN_52837 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline